Learn More
3-Hydroxy-6-methyl-2-nitropyridine, 99%
Product Code. 11357126
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357126 50 g 50g
11347126 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11357126 Supplier Thermo Scientific Alfa Aesar Supplier No. L07074.18
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 15128-90-2 | C6H6N2O3 | 154.125 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15128-90-2
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00006259
InChI Key WZMGQHIBXUAYGS-UHFFFAOYSA-N
PubChem CID 84801
SMILES CC1=NC(=C(C=C1)O)[N+](=O)[O-]
Specifications
Melting Point 105°C to 108°C
Quantity 50 g
Beilstein 974751
Synonym 3-hydroxy-6-methyl-2-nitropyridine, 6-methyl-2-nitropyridin-3-ol, 6-methyl-2-nitro-3-pyridinol, 3-pyridinol, 6-methyl-2-nitro, 3-hydroxy-2-nitro-6-picoline 3-hydroxy-6-methyl-2-nitropyridine, pubchem6630, 2-nitro-6-methylpyridin-3-ol, 3-pyridinol,6-methyl-2-nitro, 6-methyl-2-nitro-pyridin-3-ol, wzmgqhibxuaygs-uhfffaoysa
Formula Weight 154.13
Percent Purity 99%
Chemical Name or Material 3-Hydroxy-6-methyl-2-nitropyridine