accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 15128-90-2

CH3OHNNOO

CAS RN 15128-90-2

IUPAC Name: 6-methyl-2-nitropyridin-3-ol
Synonyms: 6-methyl-2-nitropyridin-3-ol
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: WZMGQHIBXUAYGS-UHFFFAOYSA-N
SMILES: CC1=CC=C(O)C(=N1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results