missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoropyridine-2-carboxylic acid, 98%

Product Code. p-7041245
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428008 1 g 1g
15438008 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428008 Supplier Thermo Scientific Alfa Aesar Supplier No. H61458.03
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 152126-31-3 | C6H4FNO2 | 141.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152126-31-3
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD04114108
InChI Key IRERRSXDWUCFIY-UHFFFAOYSA-N
Synonym 3-fluoropicolinic acid, 3-fluoro-2-pyridinecarboxylic acid, 3-fluoro-2-carboxypyridine, 2-pyridinecarboxylic acid, 3-fluoro, 2-pyridinecarboxylicacid, 3-fluoro, 3-fluoro-pyridine-2-carboxylic acid, 3-fluoropyridine-2-carboxylicacid, pubchem5140, acmc-1c4oi, 3-fluoropyridine-carboxylic
PubChem CID 2762786
IUPAC Name 3-fluoropyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)F

Specifications

Quantity 1 g
Formula Weight 141.1
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.