accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 152126-31-3

NOOHF

CAS RN 152126-31-3

IUPAC Name: 3-fluoropyridine-2-carboxylic acid
Synonyms: 3-fluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 141.1
Molecular Formula: C6H4FNO2
InChi Key: IRERRSXDWUCFIY-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results