Learn More
3-Fluoropyridine-2-carboxylic acid, 98%
Product Code. 15438008
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438008 5 g 5g
15428008 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15438008 Supplier Thermo Scientific Alfa Aesar Supplier No. H61458.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 152126-31-3 | C6H4FNO2 | 141.101 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 152126-31-3
Molecular Formula C6H4FNO2
Molecular Weight (g/mol) 141.101
MDL Number MFCD04114108
InChI Key IRERRSXDWUCFIY-UHFFFAOYSA-N
Synonym 3-fluoropicolinic acid, 3-fluoro-2-pyridinecarboxylic acid, 3-fluoro-2-carboxypyridine, 2-pyridinecarboxylic acid, 3-fluoro, 2-pyridinecarboxylicacid, 3-fluoro, 3-fluoro-pyridine-2-carboxylic acid, 3-fluoropyridine-2-carboxylicacid, pubchem5140, acmc-1c4oi, 3-fluoropyridine-carboxylic
PubChem CID 2762786
IUPAC Name 3-fluoropyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)F
Specifications
Quantity 5 g
Formula Weight 141.1
Percent Purity 98%
Chemical Name or Material 3-Fluoropyridine-2-carboxylic acid