Learn More
3-Decanol, 97%
Product Code. 11421157
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11421157
Brand: Thermo Scientific Alfa Aesar B24141.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 1565-81-7 | C10H22O | 158.285 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1565-81-7
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
MDL Number MFCD00021956
InChI Key ICEQLCZWZXUUIJ-UHFFFAOYSA-N
Synonym 3-decanol, heptyl ethyl carbinol, ethyl heptyl carbinol, 3-decanol, r, 3-decanol, s, fema no. 3605, 3-decyl alcohol, ethylheptylcarbinol, deca-8-yl alcohol, acmc-20mxdt
PubChem CID 519158
IUPAC Name decan-3-ol
SMILES CCCCCCCC(CC)O
Specifications
Density 0.83
Boiling Point 213°C
Flash Point 99°C (210°F)
Refractive Index 1.435
Quantity 25 g
Beilstein 1719658
Formula Weight 158.29
Percent Purity 97%
Chemical Name or Material 3-Decanol