accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1565-81-7

H3COHCH3

CAS RN 1565-81-7

IUPAC Name: decan-3-ol
Synonyms: decan-3-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: ICEQLCZWZXUUIJ-UHFFFAOYNA-N
SMILES: CCCCCCCC(O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results