Learn More
3-Cyclopentene-1-carboxylic acid, 95%, Thermo Scientific™
Product Code. 11497136
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497136 5 g 5g
11487136 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11497136 Supplier Thermo Scientific Alfa Aesar Supplier No. B25456.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
3-Cyclopentene-1-carboxylic acid is used as an intermediate in the production of dolasetron mesylate.
Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7686-77-3
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00101764
InChI Key XVSYDLITVYBCBD-UHFFFAOYSA-N
Synonym 3-cyclopentene-1-carboxylic acid, cyclopent-3-enecarboxylic acid, 3-cyclopentenecarboxylic acid, 3-cyclopentene-1-carhoxylic acid, cyclopent-3-ene carboxylic acid, cyclopentene-3-carboxylic acid, ghl.pd_mitscher_leg0.243, 3-cyclopentene-1-carboxylicacid, acmc-1bh22, ksc377a8b
PubChem CID 853756
IUPAC Name cyclopent-3-ene-1-carboxylic acid
SMILES C1C=CCC1C(=O)O
Specifications
Density 1.084
Boiling Point 215°C
Flash Point >110°C (230°F)
Refractive Index 1.469
Quantity 5 g
Formula Weight 112.13
Percent Purity 95%
Chemical Name or Material 3-Cyclopentene-1-carboxylic acid