CAS RN 7686-77-3
CAS RN 7686-77-3
IUPAC Name:
cyclopent-3-ene-1-carboxylate
Synonyms:
cyclopent-3-ene-1-carboxylate
Molecular Weight (g/mol):
111.12
Molecular Formula:
C6H7O2
InChi Key:
XVSYDLITVYBCBD-UHFFFAOYSA-M
SMILES:
[O-]C(=O)C1CC=CC1
1
–
1
of
1
results
1
3-Cyclopentene-1-carboxylic acid, 95%, Thermo Scientific™
CAS: 7686-77-3 Molecular Formula: C6H8O2 Molecular Weight (g/mol): 112.128 MDL Number: MFCD00101764 InChI Key: XVSYDLITVYBCBD-UHFFFAOYSA-N Synonym: 3-cyclopentene-1-carboxylic acid,cyclopent-3-enecarboxylic acid,3-cyclopentenecarboxylic acid,3-cyclopentene-1-carhoxylic acid,cyclopent-3-ene carboxylic acid,cyclopentene-3-carboxylic acid,ghl.pd_mitscher_leg0.243,3-cyclopentene-1-carboxylicacid,acmc-1bh22,ksc377a8b PubChem CID: 853756 IUPAC Name: cyclopent-3-ene-1-carboxylic acid SMILES: C1C=CCC1C(=O)O