missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline hydrochloride, 97%, Thermo Scientific™

Product Code. p-7046217
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383097 25 g 25g
11393097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11383097 Supplier Thermo Scientific Alfa Aesar Supplier No. L13484.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-85-5
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00035485
InChI Key NMGHWHCTRGZZOP-UHFFFAOYSA-N
Synonym 3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
PubChem CID 67336
IUPAC Name 3-chloroaniline;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)N.Cl

Specifications

Melting Point ∽222°C (decomposition)
Odor Characteristic
Quantity 25 g
UN Number UN2018
Beilstein 3623168
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 3-Chloroaniline hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.