accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 141-85-5

IUPAC Name: hydrogen 3-chloroaniline chloride
Synonyms: hydrogen 3-chloroaniline chloride
Molecular Weight (g/mol): 164.03
Molecular Formula: C6H7Cl2N
InChi Key: NMGHWHCTRGZZOP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results