missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-5'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 11405790
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11405790 10 g 10g
11495780 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405790 Supplier Thermo Scientific Alfa Aesar Supplier No. H26450.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886497-11-6
Molecular Formula C9H6ClF3O
Molecular Weight (g/mol) 222.591
MDL Number MFCD04115856
InChI Key HRGMIFUPRXVREB-UHFFFAOYSA-N
Synonym 3'-chloro-5'-trifluoromethyl acetophenone, 1-3-chloro-5-trifluoromethyl phenyl ethanone, 3'-chloro-5'-trifluoromethylacetophenone, 1-acetyl-5-chloro-3-trifluoromethyl benzene, ethanone,1-3-chloro-5-trifluoromethyl phenyl, 1-3-chloro-5-trifluoromethyl phenyl-ethanone, 1-3-chloro-5-trifluoromethyl phenyl ethan-1-one, ethanone, 1-3-chloro-5-trifluoromethyl phenyl
PubChem CID 3859062
IUPAC Name 1-[3-chloro-5-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=CC(=CC(=C1)Cl)C(F)(F)F

Specifications

Refractive Index 1.4825
Quantity 10 g
Formula Weight 222.6
Percent Purity 97%
Chemical Name or Material 3'-Chloro-5'-(trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.