accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 886497-11-6

H3COClFFF

CAS RN 886497-11-6

IUPAC Name: 1-[3-chloro-5-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[3-chloro-5-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 222.59
Molecular Formula: C9H6ClF3O
InChi Key: HRGMIFUPRXVREB-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(=CC(Cl)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results