Learn More
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Product Code. 11466736
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466736 1 g 1g
11476736 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11466736 Supplier Thermo Scientific Alfa Aesar Supplier No. B23889.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 261763-02-4
Molecular Formula C8H3ClF4O
Molecular Weight (g/mol) 226.555
MDL Number MFCD01631510
InChI Key MSZTVIFIFJCNRQ-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-5-trifluoromethyl benzaldehyde, benzaldehyde, 3-chloro-2-fluoro-5-trifluoromethyl, 3-chloro-2-fluoro-5-trifluoromethyl-benzaldehyde, pubchem2887, acmc-20an7f, 3-chloro-5-trifluoromethyl-2-fluorobenzaldehyde, 3-chloranyl-2-fluoranyl-5-trifluoromethyl benzaldehyde
PubChem CID 736336
IUPAC Name 3-chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
SMILES C1=C(C=C(C(=C1Cl)F)C=O)C(F)(F)F
Specifications
Density 1.54
Boiling Point 196°C
Flash Point 96°C (204°F)
Refractive Index 1.477
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde