accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 261763-02-4

FClOFFF

CAS RN 261763-02-4

IUPAC Name: 3-chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
Synonyms: 3-chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 226.55
Molecular Formula: C8H3ClF4O
InChi Key: MSZTVIFIFJCNRQ-UHFFFAOYSA-N
SMILES: FC1=C(Cl)C=C(C=C1C=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results