missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2,6-difluorotoluene, 99%, Thermo Scientific™

Product Code. 11386529
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11386529

Brand: Thermo Scientific Alfa Aesar H26159.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 221220-97-9
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD03412221
InChI Key AISIRIFPPGLTCY-UHFFFAOYSA-N
Synonym 3-bromo-2,6-difluorotoluene, 2,4-difluoro-3-methylbromobenzene, 1-bromo-2,4-difluoro-3-methyl-benzene, benzene, 1-bromo-2,4-difluoro-3-methyl, acmc-209fsf, ksc494q0f, 3-methyl-2,4-difluoro-bromobenzene, bromo-2,4-difluoro-3-methyl-benzene, 4-bromo-1,3-difluoro-2-methylbenzene
PubChem CID 11287226
IUPAC Name 1-bromo-2,4-difluoro-3-methylbenzene
SMILES CC1=C(F)C=CC(Br)=C1F

Specifications

Refractive Index 1.51
Quantity 1 g
Formula Weight 207.02
Percent Purity 99%
Chemical Name or Material 3-Bromo-2,6-difluorotoluene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis