accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 221220-97-9

CH3FBrF

CAS RN 221220-97-9

IUPAC Name: 1-bromo-2,4-difluoro-3-methylbenzene
Synonyms: 1-bromo-2,4-difluoro-3-methylbenzene
Molecular Weight (g/mol): 207.02
Molecular Formula: C7H5BrF2
InChi Key: AISIRIFPPGLTCY-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(Br)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results