Learn More
(3-Benzoylphenyl)acetic Acid, Mikromol™
Description
(3-Benzoylphenyl)acetic Acid
Specifications
Specifications
| Chemical Name or Material | (3-Benzoylphenyl)acetic Acid |
| CAS | 22071-22-3 |
| Analyte or Component Names | (3-Benzoylphenyl)acetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H12 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Ketoprofen Imp. B (EP), (3-Benzoylphenyl)acetic Acid, 2-(3-Benzoylphenyl)acetic acid, RU 4462, m-Benzoylphenylacetic acid, Ketoprofen Impurity B |
| InChI Formula | InChI=1S/C15H12O3/c16-14(17)10-11-5-4-8-13(9-11)15(18)12-6-2-1-3-7-12/h1-9H,10H2,(H,16,17) |
| SMILES | OC(=O)Cc1cccc(c1)C(=O)c2ccccc2 |
| IUPAC Name | 2-(3-benzoylphenyl)acetic acid |
| Show More |