Learn More
3-Amino-4-phenoxy-5-sulfamoylbenzoic Acid, Mikromol™
Description
3-Amino-4-phenoxy-5-sulfamoylbenzoic Acid
Specifications
Specifications
| Chemical Name or Material | Desbutyl Bumetanide |
| CAS | 28328-54-3 |
| Analyte or Component Names | Desbutyl Bumetanide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H12 N2 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Bumetanide Imp. B (EP), Bumetanide USP Related Compound A, Bumetanide USP RC A, Benzoic acid, 3-amino-5-(aminosulfonyl)-4-phenoxy-, Benzoic acid, 3-amino-4-phenoxy-5-sulfamoyl- (8CI), 3-Amino-4-phenoxy-5-sulfamoylbenzoic acid, Desbutylbumetanide, PF 1578, Bumetanide Related Compound A, Bumetanide Impurity B |
| InChI Formula | InChI=1S/C13H12N2O5S/c14-10-6-8(13(16)17)7-11(21(15,18)19)12(10)20-9-4-2-1-3-5-9/h1-7H,14H2,(H,16,17)(H2,15,18,19) |
| SMILES | Nc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O |
| IUPAC Name | 3-amino-4-phenoxy-5-sulfamoylbenzoic acid |
| Show More |