Learn More
3-Amino-4-phenoxy-5-sulfamoylbenzoic Acid, Mikromol™
Product Code. 17932756
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932756 25 mg 25mg
17972856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17932756 Supplier Mikromol Supplier No. MM1290.020025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Amino-4-phenoxy-5-sulfamoylbenzoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Desbutyl Bumetanide
CAS 28328-54-3
Analyte or Component Names Desbutyl Bumetanide
Shipping Condition Room Temperature
Molecular Formula C13 H12 N2 O5 S
Certifications/Compliance ISO 17025
Synonym Bumetanide Imp. B (EP), Bumetanide USP Related Compound A, Bumetanide USP RC A, Benzoic acid, 3-amino-5-(aminosulfonyl)-4-phenoxy-, Benzoic acid, 3-amino-4-phenoxy-5-sulfamoyl- (8CI), 3-Amino-4-phenoxy-5-sulfamoylbenzoic acid, Desbutylbumetanide, PF 1578, Bumetanide Related Compound A, Bumetanide Impurity B
InChI Formula InChI=1S/C13H12N2O5S/c14-10-6-8(13(16)17)7-11(21(15,18)19)12(10)20-9-4-2-1-3-5-9/h1-7H,14H2,(H,16,17)(H2,15,18,19)
SMILES Nc1cc(cc(c1Oc2ccccc2)S(=O)(=O)N)C(=O)O
IUPAC Name 3-amino-4-phenoxy-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 308.31
Quantity 25 mg
Formula Weight 308.0467 g/mol