Learn More
3-Amino-2-methyl-6-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Product Code. 15426457
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426457 1 g 1g
15438545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15426457 Supplier Thermo Scientific Alfa Aesar Supplier No. H33609.03
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 383907-17-3
Molecular Formula C7H7F3N2
Molecular Weight (g/mol) 176.14
MDL Number MFCD01311991
InChI Key FUJKUIAVFVGTGS-UHFFFAOYSA-N
PubChem CID 3521850
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridin-3-amine
SMILES CC1=C(N)C=CC(=N1)C(F)(F)F
Specifications
Melting Point 106°C to 108°C
Quantity 1 g
Synonym 2-methyl-6-trifluoromethyl pyridin-3-amine, 2-methyl-6-trifluoromethyl-3-pyridinamine, 3-amino-2-methyl-6-trifluoromethyl pyridine, 2-methyl-6-trifluoromethylpyridin-3-yl amine, 2-methyl-6-trifluoromethyl-pyridin-3-ylamine, 2-methyl-6-trifluoromethyl-3-pyridylamine, 3-pyridinamine,2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethyl pyridine-3-amine, 3-pyridinamine, 2-methyl-6-trifluoromethyl
Solubility Information Slightly soluble in water.
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methyl-6-(trifluoromethyl)pyridine