accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 383907-17-3

CH3NFFFNH2

CAS RN 383907-17-3

IUPAC Name: 2-methyl-6-(trifluoromethyl)pyridin-3-amine
Synonyms: 2-methyl-6-(trifluoromethyl)pyridin-3-amine
Molecular Weight (g/mol): 176.14
Molecular Formula: C7H7F3N2
InChi Key: FUJKUIAVFVGTGS-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC(=N1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)