Learn More
3’-Acetyl-4’-(2,3-epoxypropoxy)butyranilide, TRC
Product Code. 30007377
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007377 100 mg 100mg
30012286 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30007377 Supplier Toronto Research Chemicals Supplier No. TRCA173780100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 28197-66-2
Molecular Formula C15 H19 N O4
Molecular Weight (g/mol) 277.32
Synonym N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide, Acebutolol Hydrochloride Imp. A (EP)
IUPAC Name N-[3-acetyl-4-(oxiran-2-ylmethoxy)phenyl]butanamide
SMILES CCCC(=O)Nc1ccc(OCC2CO2)c(c1)C(=O)C
Specifications
Analyte or Component Names N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C15H19NO4/c1-3-4-15(18)16-11-5-6-14(13(7-11)10(2)17)20-9-12-8-19-12/h5-7,12H,3-4,8-9H2,1-2H3,(H,16,18)
Solution Type Neat
Industry Type Pharmaceutical
Quantity 100 mg
Formula Weight 277.1314
Percent Purity >95%
Chemical Name or Material N-[3-Acetyl-4-[(2RS)-oxiran-2-ylmethoxy]phenyl]butanamide
Grade HPLC