missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dihydroxyflavone, 97%

Product Code. p-7029518
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481620 1 g 1g
11491620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481620 Supplier Thermo Scientific Alfa Aesar Supplier No. H26989.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 108238-41-1 | C15H10O4 | 254.241 g/mol

3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,6-Dihydroxyflavone,is used as an oxidative stress inducer - apoptosis inducer, it is widely used as a pharmaceutical intermediate and in cell survival applications.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108238-41-1
Molecular Formula C15H10O4
Molecular Weight (g/mol) 254.241
MDL Number MFCD00143082
InChI Key XHLOLFKZCUCROE-UHFFFAOYSA-N
Synonym 3,6-dihydroxyflavone, 3,6-dihydroxy-2-phenyl-4h-chromen-4-one, 6-hydroxyflavonol, 3,6-dihydroxy-flavone, 4h-1-benzopyran-4-one, 3,6-dihydroxy-2-phenyl, acmc-1c7fz, 3,6-dihydroxy-2-phenylchromone
PubChem CID 688659
IUPAC Name 3,6-dihydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=C(C(=O)C3=C(O2)C=CC(=C3)O)O

Specifications

Melting Point 231°C to 234°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 254.24
Percent Purity 97%
Chemical Name or Material 3,6-Dihydroxyflavone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.