accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 108238-41-1

OOOHHO

CAS RN 108238-41-1

IUPAC Name: 3,6-dihydroxy-2-phenyl-4H-chromen-4-one
Synonyms: 3,6-dihydroxy-2-phenylchromen-4-one
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: XHLOLFKZCUCROE-UHFFFAOYSA-N
SMILES: OC1=CC=C2OC(=C(O)C(=O)C2=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results