Learn More
3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine), Mikromol™
Product Code. 17912286
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 17912286
Brand: Mikromol MM0077.05
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine)
CAS 303-54-8
Analyte or Component Names 3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine (Depramine; 10,11-Dehydroimipramine)
Shipping Condition Room Temperature
Molecular Formula C19 H22 N2
Certifications/Compliance ISO 17025
Synonym Imipramine Hydrochloride Imp. B (EP), Imipramine Imp. B (EP), Depramine, 3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine, 10,11-Dehydroimipramine, 5H-Dibenz[b,f]azepine-5-propanamine, N,N-dimethyl-, 5H-Dibenz[b,f]azepine, 5-[3-(dimethylamino)propyl]- (6CI,7CI,8CI), N,N-Dimethyl-5H-dibenz[b,f]azepine-5-propanamine, 5-[3-(Dimethylamino)propyl]-5H-dibenz[b,f]azepine, Balipramine, G 31406, GP 31406, Imipramine Hydrochloride Impurity B, Imipramine Impurity B
InChI Formula InChI=1S/C19H22N2/c1-20(2)14-7-15-21-18-10-5-3-8-16(18)12-13-17-9-4-6-11-19(17)21/h3-6,8-13H,7,14-15H2,1-2H3
SMILES CN(C)CCCN1c2ccccc2C=Cc3ccccc13
IUPAC Name 3-benzo[b][1]benzazepin-11-yl-N,N-dimethylpropan-1-amine
Molecular Weight (g/mol) 278.39
Quantity 100 mg
Formula Weight 278.1783 g/mol