Learn More
3,4-Dimethoxy-2-methylpyridine N-Oxide, Mikromol
Product Code. 30460454
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460454 4 x 25 mg 100mg
30049970 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30460454 Supplier Mikromol Supplier No. MM0099.1500254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3,4-Dimethoxy-2-methylpyridine N-Oxide, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3,4-Dimethoxy-2-methylpyridine N-Oxide
CAS 72830-07-0
Analyte or Component Names 3,4-Dimethoxy-2-methylpyridine N-Oxide
Shipping Condition Room Temperature
Molecular Formula C8H11NO3
Certifications/Compliance ISO 9001
Synonym Pyridine, 3,4-dimethoxy-2-methyl-, 1-oxide, 2-Methyl-3,4-dimethoxypyridine N-oxide, 3,4-Dimethoxy-2-methylpyridine 1-oxide, 3,4-Dimethoxy-2-methylpyridine-N-oxide
InChI Formula InChI=1S/C8H11NO3/c1-6-8(12-3)7(11-2)4-5-9(6)10/h4-5H,1-3H3
Solution Type Neat
SMILES COc1cc[n+]([O-])c(C)c1OC
IUPAC Name 3,4-dimethoxy-2-methyl-1-oxido-pyridin-1-ium
Molecular Weight (g/mol) 169.18
Quantity 4 x 25 mg
Formula Weight 169.0739 g/mol