Learn More
3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™
Product Code. 11302676
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302676 5 g 5g
11312676 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11302676 Supplier Thermo Scientific Alfa Aesar Supplier No. L08717.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5409-31-4
Molecular Formula C11H14O4
Molecular Weight (g/mol) 210.229
MDL Number MFCD00002504
InChI Key VVKCVAPLTRZJHH-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-diethoxy, benzoicacid34diethoxy, 3,4-diethoxy-benzoic acid, acmc-1apyn, ksc271i5p, 3,4-bis ethyloxy benzoic acid, 3,4-diethoxybenzoic acid
PubChem CID 79417
IUPAC Name 3,4-diethoxybenzoic acid
SMILES CCOC1=C(C=C(C=C1)C(=O)O)OCC
Specifications
Melting Point 167°C to 170°C
Quantity 5 g
Beilstein 2651892
Formula Weight 210.23
Percent Purity 99%
Chemical Name or Material 3,4-Diethoxybenzoic acid