CAS RN 5409-31-4
CAS RN 5409-31-4
IUPAC Name:
3,4-diethoxybenzoic acid
Synonyms:
BENZOICACID34diethoxy
Molecular Weight (g/mol):
210.23
Molecular Formula:
C11H14O4
InChi Key:
VVKCVAPLTRZJHH-UHFFFAOYSA-N
SMILES:
CCOC1=CC=C(C=C1OCC)C(O)=O
1
–
2
of
2
results
2
3,4-Diethoxybenzoic acid, 99%, Thermo Scientific™
CAS: 5409-31-4 Molecular Formula: C11H14O4 Molecular Weight (g/mol): 210.229 MDL Number: MFCD00002504 InChI Key: VVKCVAPLTRZJHH-UHFFFAOYSA-N Synonym: benzoic acid, 3,4-diethoxy,benzoicacid34diethoxy,3,4-diethoxy-benzoic acid,acmc-1apyn,ksc271i5p,3,4-bis ethyloxy benzoic acid,3,4-diethoxybenzoic acid PubChem CID: 79417 IUPAC Name: 3,4-diethoxybenzoic acid SMILES: CCOC1=C(C=C(C=C1)C(=O)O)OCC