missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromobenzoyl)propionic acid, 98+%, Thermo Scientific™

Product Code. p-7044005
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356746 1 g 1g
11366746 5 g 5g
11376746 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356746 Supplier Thermo Scientific Alfa Aesar Supplier No. L04008.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6340-79-0
Molecular Formula C10H9BrO3
Molecular Weight (g/mol) 257.08
MDL Number MFCD00016563
InChI Key ZODFRCZNTXLDDW-UHFFFAOYSA-N
Synonym 3-4-bromobenzoyl propionic acid, 4-4-bromophenyl-4-oxobutanoic acid, 3-p-bromobenzoyl propionic acid, 4-4-bromo-phenyl-4-oxo-butyric acid, 3-4-bromobenzoyl propanoic acid, 4-4-bromophenyl-4-oxobutyric acid, 4-4-bromophenyl-4-oxo-butyric acid, 4-4-bromophenyl-4-oxo-butanoic acid, benzenebutanoic acid, 4-bromo-gamma-oxo, 4-bromo-gamma-oxo-benzenebutanoic acid
PubChem CID 80646
IUPAC Name 4-(4-bromophenyl)-4-oxobutanoic acid
SMILES OC(=O)CCC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 145°C to 148°C
Quantity 1 g
Beilstein 2104410
Formula Weight 257.09
Percent Purity ≥98%
Chemical Name or Material 3-(4-Bromobenzoyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.