CAS RN 6340-79-0
IUPAC Name:
4-(4-bromophenyl)-4-oxobutanoic acid
Synonyms:
4-(4-bromophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol):
257.08
Molecular Formula:
C10H9BrO3
InChi Key:
ZODFRCZNTXLDDW-UHFFFAOYSA-N
SMILES:
OC(=O)CCC(=O)C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
3-(4-Bromobenzoyl)propionic acid, 98+%, Thermo Scientific™
CAS: 6340-79-0 Molecular Formula: C10H9BrO3 Molecular Weight (g/mol): 257.08 MDL Number: MFCD00016563 InChI Key: ZODFRCZNTXLDDW-UHFFFAOYSA-N Synonym: 3-4-bromobenzoyl propionic acid,4-4-bromophenyl-4-oxobutanoic acid,3-p-bromobenzoyl propionic acid,4-4-bromo-phenyl-4-oxo-butyric acid,3-4-bromobenzoyl propanoic acid,4-4-bromophenyl-4-oxobutyric acid,4-4-bromophenyl-4-oxo-butyric acid,4-4-bromophenyl-4-oxo-butanoic acid,benzenebutanoic acid, 4-bromo-gamma-oxo,4-bromo-gamma-oxo-benzenebutanoic acid PubChem CID: 80646 IUPAC Name: 4-(4-bromophenyl)-4-oxobutanoic acid SMILES: OC(=O)CCC(=O)C1=CC=C(Br)C=C1