Learn More
3-[4-(3-Chlorophenyl)piperazin-1-yl]propanol, Mikromol™
Product Code. 30051263
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051263 100 mg 100mg
30049571 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051263 Supplier Mikromol Supplier No. MM0421.10
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[4-(3-Chlorophenyl)piperazin-1-yl]propanol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[4-(3-Chlorophenyl)piperazin-1-yl]propanol
CAS 32229-98-4
Analyte or Component Names 3-[4-(3-Chlorophenyl)piperazin-1-yl]propanol
Shipping Condition Room Temperature
Molecular Formula C13 H19 Cl N2 O
Certifications/Compliance ISO 17025
Synonym 3-[4-(3-Chlorophenyl)piperazin-1-yl]propanol
InChI Formula InChI=1S/C13H19ClN2O/c14-12-3-1-4-13(11-12)16-8-6-15(7-9-16)5-2-10-17/h1,3-4,11,17H,2,5-10H2
SMILES OCCCN1CCN(CC1)c2cccc(Cl)c2
IUPAC Name 3-[4-(3-chlorophenyl)piperazin-1-yl]propan-1-ol
Molecular Weight (g/mol) 254.76
Quantity 100 mg
Formula Weight 254.1186 g/mol