Learn More
3,3'-(Isopropylazanediyl)bis(1-(4-((2-isopropoxyethoxy)methyl)phenoxy)propan-2-ol), TRC
Specifications
Specifications
| Chemical Name or Material | 1,1'-[(1-Methylethyl)imino]bis(1-(4-(2-isopropoxyethoxy)methyl)phenoxy)propan-2-ol |
| CAS | 2775292-22-1 |
| Analyte or Component Names | 1,1'-[(1-Methylethyl)imino]bis(1-(4-(2-isopropoxyethoxy)methyl)phenoxy)propan-2-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H53 N O8 |
| Synonym | 1-[[2-Hydroxy-3-[4-(2-isopropoxyethoxymethyl)phenoxy]propyl]-isopropyl-amino]-3-[4-(2-isopropoxyethoxymethyl)phenoxy]propan-2-ol, 3,3'-(Isopropylazanediyl)bis(1-(4-((2-isopropoxyethoxy)methyl)phenoxy)propan-2-ol, N,N-bis-[2-Hydroxy-3-[4-(2-isopropoxyethoxymethyl)phenoxy]propyl](isopropyl)amine, 1,1'-[(1-Methylethyl)imino]bis(1-(4-((2-isopropoxyethoxy)methyl)phenoxy)propan-2-ol |
| InChI Formula | InChI=1S/C33H53NO8/c1-25(2)34(19-30(35)23-41-32-11-7-28(8-12-32)21-37-15-17-39-26(3)4)20-31(36)24-42-33-13-9-29(10-14-33)22-38-16-18-40-27(5)6/h7-14,25-27,30-31,35-36H,15-24H2,1-6H3 |
| Solution Type | Neat |
| SMILES | CC(C)OCCOCc1ccc(OCC(O)CN(CC(O)COc2ccc(COCCOC(C)C)cc2)C(C)C)cc1 |
| IUPAC Name | 1-[[2-hydroxy-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propyl]-propan-2-ylamino]-3-[4-(2-propan-2-yloxyethoxymethyl)phenoxy]propan-2-ol |
| Show More |