Learn More
3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound), Mikromol™
Product Code. 30051551
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051551 25 mg 25mg
30049472 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30051551 Supplier Mikromol Supplier No. MM0128.040025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound)
CAS 85045-98-3
Analyte or Component Names 3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea (Bromhydrin Compound)
Shipping Condition Room Temperature
Molecular Formula C16 H23 Br N2 O4
Certifications/Compliance ISO 9001
Synonym Celiprolol Hydrochloride Imp. H (EP), Celiprolol Imp. H (EP), 3-[3-Acetyl-4-[(2RS)-3-bromo-2-hydroxypropoxy]phenyl]-1,1-diethylurea, Bromhydrin Compound of Celiprolol, Celiprolol Hydrochloride Impurity H, Celiprolol Impurity H
InChI Formula InChI=1S/C16H23BrN2O4/c1-4-19(5-2)16(22)18-12-6-7-15(14(8-12)11(3)20)23-10-13(21)9-17/h6-8,13,21H,4-5,9-10H2,1-3H3,(H,18,22)
SMILES CCN(CC)C(=O)Nc1ccc(OCC(O)CBr)c(c1)C(=O)C
IUPAC Name 3-[3-acetyl-4-(3-bromo-2-hydroxypropoxy)phenyl]-1,1-diethylurea
Molecular Weight (g/mol) 387.27
Quantity 25 mg
Formula Weight 386.0841 g/mol