missing translation for 'onlineSavingsMsg'
Learn More

3,3',4,4'-Tetrachlorobiphenyl, TRC

Product Code. 30455776
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30455776 5 mg -
30452239 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455776 Supplier Toronto Research Chemicals Supplier No. TRCT8899755MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32598-13-3
Molecular Formula C12 H6 Cl4
Molecular Weight (g/mol) 291.99
Synonym 1,1'-Biphenyl, 3,3',4,4'-tetrachloro-, Biphenyl, 3,3',4,4'-tetrachloro- (8CI), 3,3',4,4'-Tetrachloro-1,1'-biphenyl, 3,3',4,4'-Tetrachlorobiphenyl, 3,4,3',4'-Tetrachlorobiphenyl, CB 77, PCB 77, Polychlorinated biphenyl-77, TeCB 77, 3,3',4,4'-TetraCB
IUPAC Name 1,2-dichloro-4-(3,4-dichlorophenyl)benzene
SMILES Clc1ccc(cc1Cl)c2ccc(Cl)c(Cl)c2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H6Cl4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H
Formula Weight 289.9224
Chemical Name or Material PCB 77 (3,3',4,4'-Tetrachlorobiphenyl)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.