Learn More
3,3',4,4'-Tetrachlorobiphenyl, TRC
Product Code. 30452239
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452239 50 mg 50mg
30455776 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30452239 Supplier Toronto Research Chemicals Supplier No. TRCT88997550MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32598-13-3
Molecular Formula C12 H6 Cl4
Molecular Weight (g/mol) 291.99
Synonym 1,1'-Biphenyl, 3,3',4,4'-tetrachloro-, Biphenyl, 3,3',4,4'-tetrachloro- (8CI), 3,3',4,4'-Tetrachloro-1,1'-biphenyl, 3,3',4,4'-Tetrachlorobiphenyl, 3,4,3',4'-Tetrachlorobiphenyl, CB 77, PCB 77, Polychlorinated biphenyl-77, TeCB 77, 3,3',4,4'-TetraCB
IUPAC Name 1,2-dichloro-4-(3,4-dichlorophenyl)benzene
SMILES Clc1ccc(cc1Cl)c2ccc(Cl)c(Cl)c2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H6Cl4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H
Formula Weight 289.9224
Chemical Name or Material PCB 77 (3,3',4,4'-Tetrachlorobiphenyl)