missing translation for 'onlineSavingsMsg'
Learn More

3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid, 97%, Thermo Scientific™

Product Code. 15466697
Change view
Click to view available options
Quantity:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
15466697 1 g 1g
15476697 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15466697 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H33829.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1373921-13-1
Molecular Formula C11H11F3O3
Molecular Weight (g/mol) 248.201
MDL Number MFCD22201060
InChI Key LNOAXHKIMRSNOJ-UHFFFAOYSA-N
Synonym 3-2-methyl-4-trifluoromethoxy phenyl propionic acid, 3-2-methyl-4-trifluoromethoxy phenyl propanoic acid
PubChem CID 86277667
IUPAC Name 3-[2-methyl-4-(trifluoromethoxy)phenyl]propanoic acid
SMILES CC1=C(C=CC(=C1)OC(F)(F)F)CCC(=O)O

Specifikationer

Quantity 1 g
Formula Weight 248.2
Percent Purity 97%
Chemical Name or Material 3-[2-Methyl-4-(trifluoromethoxy)phenyl]propionic acid
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.