accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1373921-13-1

CH3OFFFOOH

CAS RN 1373921-13-1

IUPAC Name: 3-[2-methyl-4-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[2-methyl-4-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 248.2
Molecular Formula: C11H11F3O3
InChi Key: LNOAXHKIMRSNOJ-UHFFFAOYSA-N
SMILES: CC1=C(CCC(O)=O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results