missing translation for 'onlineSavingsMsg'
Learn More

3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid, 97%, Thermo Scientific™

Product Code. 15458135
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458135 250 mg 250mg
15448135 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458135 Supplier Thermo Scientific Alfa Aesar Supplier No. H32595.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as intermediates. Also as laboratory reagents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1092460-63-3
Molecular Formula C11H8F6O2
Molecular Weight (g/mol) 286.17
MDL Number MFCD11519312
InChI Key POPIIFPERMLVHR-UHFFFAOYSA-N
Synonym 3-2,4-bis trifluoromethyl phenyl propionic acid, 3-2,4-bis trifluoromethyl phenyl propanoic acid, 2,4-bis trifluoromethyl phenyl propanoic acid, 3-2,4-bis trifluoromethyl phenyl propionic acid, jrd
PubChem CID 54772349
IUPAC Name 3-[2,4-bis(trifluoromethyl)phenyl]propanoic acid
SMILES OC(=O)CCC1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 74°C to 79°C
Quantity 250 mg
Formula Weight 286.17
Percent Purity 97%
Chemical Name or Material 3-[2,4-Bis(trifluoromethyl)phenyl]propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.