accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1092460-63-3

IUPAC Name: 3-[2,4-bis(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[2,4-bis(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 286.17
Molecular Formula: C11H8F6O2
InChi Key: POPIIFPERMLVHR-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results