Learn More
(3β)-17-Iodoandrosta-5,16-dien-3-ol, Mikromol
Description
(3β)-17-Iodoandrosta-5,16-dien-3-ol, Mikromol
Specifications
Specifications
| Chemical Name or Material | 17-Iodo-androsta-5,16-diene-3beta-ol |
| CAS | 32138-69-5 |
| Analyte or Component Names | 17-Iodo-androsta-5,16-diene-3beta-ol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19H27IO |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3β)-17-Iodo-androsta-5,16-dien-3-ol |
| Solution Type | Neat |
| SMILES | OC[C@]1(O)C(O)C(O)[C@@H](O)CO1 |
| Molecular Weight (g/mol) | 398.32 |
| Show More |