Learn More
3-(1-Methoxyethyl)benzophenone, Mikromol™
Product Code. 17922186
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17922186 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17922186 Supplier Mikromol Supplier No. MM0001.17
Please to purchase this item. Need a web account? Register with us today!
Denna artikel kan inte returneras. Se returpolicy
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(1-Methoxyethyl)benzophenone
TRUSTED_SUSTAINABILITY
Specifikationer
Chemical Name or Material 3-(1-Methoxyethyl)benzophenone
CAS 2731671-45-5
Analyte or Component Names 3-(1-Methoxyethyl)benzophenone
Shipping Condition Room Temperature
Molecular Formula C16 H16 O2
Certifications/Compliance ISO 9001
Synonym Oxothiamine, 3-(1-Methoxyethyl)benzophenone
InChI Formula InChI=1S/C16H16O2/c1-12(18-2)14-9-6-10-15(11-14)16(17)13-7-4-3-5-8-13/h3-12H,1-2H3
SMILES COC(C)c1cccc(c1)C(=O)c2ccccc2
IUPAC Name [3-(1-methoxyethyl)phenyl]-phenylmethanone
Molecular Weight (g/mol) 240.3
Quantity 100 mg
Formula Weight 240.115 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.