Learn More
(2S,5R,6R)-6-[[(2S)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Amoxicillin), Mikromol™
Description
(2S,5R,6R)-6-[[(2S)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid (L-Amoxicillin)
Specifications
Specifications
| Chemical Name or Material | L-Amoxicillin |
| CAS | 26889-93-0 |
| Analyte or Component Names | L-Amoxicillin |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C16 H19 N3 O5 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Amoxicillin Trihydrate Imp. B (EP), Amoxicillin Imp. B (EP), Amoxicillin USP Related Compound B, Amoxicillin USP RC B, (2S,5R,6R)-6-[[(2S)-2-Amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic Acid, L-Amoxicillin, Amoxicillin Related Compound B, Amoxicillin Trihydrate Impurity B, Amoxicillin Impurity B |
| InChI Formula | InChI=1S/C16H19N3O5S/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24)/t9-,10+,11-,14+/m0/s1 |
| SMILES | CC1(C)S[C@@H]2[C@H](NC(=O)[C@@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(=O)O |
| IUPAC Name | (2S,5R,6R)-6-[[(2S)-2-amino-2-(4-hydroxyphenyl)acetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid |
| Show More |