Learn More
(2RS)-N-Methyl-2-(10H-phenothiazin-10-yl)propan-1-amine Hydrochloride (Norisopromethazine Hydrochloride), Mikromol™
Product Code. 30052324
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052324 100 mg 100mg
30051216 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30052324 Supplier Mikromol Supplier No. MM0412.06
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-N-Methyl-2-(10H-phenothiazin-10-yl)propan-1-amine Hydrochloride (Norisopromethazine Hydrochloride)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Norisopromethazine Hydrochloride
CAS 2726492-60-8
Analyte or Component Names Norisopromethazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H18 N2 S . Cl H
Certifications/Compliance ISO 17025
Synonym (2RS)-N-Methyl-2-(10H-phenothiazin-10-yl)propan-1-amine Hydrochloride, Norisopromethazine Hydrochloride
InChI Formula InChI=1S/C16H18N2S.ClH/c1-12(11-17-2)18-13-7-3-5-9-15(13)19-16-10-6-4-8-14(16)18;/h3-10,12,17H,11H2,1-2H3;1H
SMILES Cl.CNCC(C)N1c2ccccc2Sc3ccccc13
IUPAC Name N-methyl-2-phenothiazin-10-ylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 306.85
Quantity 100 mg
Formula Weight 306.0957 g/mol