Learn More
(2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine (Dehydrotrimipramine), Mikromol™
Description
(2RS)-3-(5H-Dibenzo[b,f]azepin-5-yl)-N,N,2-trimethylpropan-1-amine (Dehydrotrimipramine)
Specifications
Specifications
| Chemical Name or Material | Dehydrotrimipramine |
| CAS | 315-69-5 |
| Analyte or Component Names | Dehydrotrimipramine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H24 N2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | N,N,β-Trimethyl-5H-dibenz[b,f]azepine-5-propanamine, 5-[3-(Dimethylamino)-2-methylpropyl]-5H-dibenz[b,f]azepine, Dehydro trimipramine, Trimipramine Related Compound A, Trimipramine Maleate Impurity C, Trimipramine Impurity C |
| InChI Formula | InChI=1S/C20H24N2/c1-16(14-21(2)3)15-22-19-10-6-4-8-17(19)12-13-18-9-5-7-11-20(18)22/h4-13,16H,14-15H2,1-3H3 |
| SMILES | CC(CN(C)C)CN1c2ccccc2C=Cc3ccccc13 |
| IUPAC Name | 3-benzo[b][1]benzazepin-11-yl-N,N,2-trimethylpropan-1-amine |
| Show More |