Learn More
(2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane, Mikromol™
Description
(2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane |
| CAS | 32778-08-8 |
| Analyte or Component Names | (2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C3 H Cl2 F5 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Isoflurane Imp. C (EP), Isoflurane USP Related Compound A, Isoflurane USP RC A, (2RS)-2-Chloro-2-(chlorodifluoromethoxy)-1,1,1-trifluoroethane, Isoflurane Related Compound A, Isoflurane Impurity C |
| InChI Formula | InChI=1S/C3HCl2F5O/c4-1(2(6,7)8)11-3(5,9)10/h1H |
| SMILES | FC(F)(F)C(Cl)OC(F)(F)Cl |
| IUPAC Name | 2-chloro-2-[chloro(difluoro)methoxy]-1,1,1-trifluoroethane |
| Show More |