Learn More
(2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride, Mikromol™
Description
(2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride |
| CAS | 2732309-77-0 |
| Analyte or Component Names | (2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H38 N2 O4 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Verapamil Hydrochloride Imp. O (EP), Verapamil Imp. O (EP), (2RS)-2-(3,4-Dimethoxyphenyl)-5-[2-[2-(3,4-dimethoxy-phenyl)ethyl](methyl)amino]- 2-propylpentane-nitrile Hydrochloride, Verapamil Hydrochloride Imp. O (EP) as Hydrochloride, Verapamil Hydrochloride Impurity O as Hydrochloride, Verapamil Impurity O as Hydrochloride |
| InChI Formula | InChI=1S/C9H10N4O2S2/c1-6-11-12-9(16-6)13-17(14,15)8-4-2-7(10)3-5-8/h2-5H,10H2,1H3,(H,12,13) |
| SMILES | Cl.CCCC(CCCN(C)CCc1ccc(OC)c(OC)c1)(C#N)c2ccc(OC)c(OC)c2 |
| IUPAC Name | 2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-methylamino]-2-propylpentanenitrile;hydrochloride |
| Show More |