Learn More
(2RS)-2-(2,6-Di-methylphenoxy)propan-1-amine Hydrochloride, Mikromol™
Description
(2RS)-2-(2,6-Di-methylphenoxy)propan-1-amine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-(2,6-Di-methylphenoxy)propan-1-amine Hydrochloride |
| CAS | 2724689-76-1 |
| Analyte or Component Names | (2RS)-2-(2,6-Di-methylphenoxy)propan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H17 N O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Mexiletine Hydrochloride Imp. D (EP), Mexiletine Imp. D (EP), (2RS)-2-(2,6-Di-methylphenoxy)propan-1-amine Hydrochloride, Mexiletine Hydrochloride Imp. D (EP) as Hydrochloride, Mexiletine Hydrochloride Impurity D as Hydrochloride, Mexiletine Impurity D as Hydrochloride |
| InChI Formula | InChI=1S/C21H25NO3.ClH/c1-2-14-22-15-18(23)16-25-21-11-7-6-10-19(21)20(24)13-12-17-8-4-3-5-9-17;/h3-13,18,22-23H,2,14-16H2,1H3;1H/b13-12+; |
| SMILES | Cl.CC(CN)Oc1c(C)cccc1C |
| IUPAC Name | 2-(2,6-dimethylphenoxy)propan-1-amine;hydrochloride |
| Show More |