Learn More
(2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethanamine Nitrate, Mikromol™
Description
(2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethanamine Nitrate
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethanamine Nitrate |
| CAS | 2724689-95-4 |
| Analyte or Component Names | (2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethanamine Nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 Cl4 N O . H N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Miconazole Nitrate Imp. C (EP), Miconazole USP Related Compound C, Miconazole USP RC C, (2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichloro-phenyl)ethanamine Nitrate, Miconazole Nitrate Imp. C (EP) as Nitrate, Miconazole Related Compound C as Nitrate, Miconazole Impurity C as Nitrate, Miconazole Nitrate Impurity C as Nitrate, Miconazole Impurity C as Nitrate |
| InChI Formula | InChI=1S/C23H32ClN3.2ClH/c1-25(2)14-6-15-26(3)16-7-17-27-22-9-5-4-8-19(22)10-11-20-12-13-21(24)18-23(20)27;;/h4-5,8-9,12-13,18H,6-7,10-11,14-17H2,1-3H3;2*1H |
| SMILES | NCC(OCc1ccc(Cl)cc1Cl)c2ccc(Cl)cc2Cl.O[N+](=O)[O-] |
| IUPAC Name | 2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethanamine;nitric acid |
| Show More |