Learn More
(2RS)-1-[4-(2-Butoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride, Mikromol™
Description
(2RS)-1-[4-(2-Butoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (2RS)-1-[4-(2-Butoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride |
| CAS | 2724726-61-6 |
| Analyte or Component Names | (2RS)-1-[4-(2-Butoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H31 N O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Betaxolol Hydrochloride Imp. E (EP), Betaxolol Imp. E (EP), (2RS)-1-[4-(2-Butoxyethyl)phenoxy]-3-[(1-methylethyl)amino]propan-2-ol Hydrochloride, Betaxolol Hydrochloride Imp. E (EP) as Hydrochloride, Betaxolol Hydrochloride Impurity E as Hydrochloride, Betaxolol Impurity E as Hydrochloride |
| InChI Formula | InChI=1S/C18H31NO3.ClH/c1-4-5-11-21-12-10-16-6-8-18(9-7-16)22-14-17(20)13-19-15(2)3;/h6-9,15,17,19-20H,4-5,10-14H2,1-3H3;1H |
| SMILES | Cl.CCCCOCCc1ccc(OCC(O)CNC(C)C)cc1 |
| IUPAC Name | 1-[4-(2-butoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride |
| Show More |